Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The specific function of the protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RP13-531C17.2; UPF0688 protein C1orf174
Gene Aliases: C1orf174
UniProt ID: (Human) Q8IYL3
Entrez Gene ID: (Human) 339448
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support