Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: EAGVSVQQGA ASLPLGGCRV VSDSRLAKTR DGLSVPKHSA GSGAEESNSS
Sequence homology: Human: 100%
The specific function of the protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RP13-531C17.2; UPF0688 protein C1orf174
Gene Aliases: C1orf174
UniProt ID: (Human) Q8IYL3
Entrez Gene ID: (Human) 339448
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support