Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YSMQKKKPRE LMNFKSFVEE LNSHHNKKHG VFSKLPRNPK TPTERIYWAN
For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
Predicted homology: Cow: 86%; Dog: 79%; Horse: 86%; Human: 100%; Mouse: 79%; Rabbit: 93%; Rat: 79%
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ciliary microtubule-associated protein 2; Lymphocyte expansion molecule; uncharacterized protein C1orf177
Gene Aliases: C1orf177; CIMAP2; LEM; LEXM
UniProt ID: (Human) Q3ZCV2
Entrez Gene ID: (Human) 163747
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support