Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MFAIQPGLA EGGQFLGDPP PGLCQPELQP DSNSNFMASA KDANENWHGM PGRVEPILRR SSSESPSDNQ AFQAPGSPEE GVRSPPEGAE IPGAEPEKMG GAGTVCSPLE DNGYASSSLS IDSRSSSPEP ACGTPRGPGP PDPLLPSVAQ AVQHLQVQER YKEQEKEKHH VHLVMYRRLA LLQWIRGLQH QLIDQQARLQ ESFDTILDNR KELIRCLQQR AAPSRPQDQA (1-229 aa encoded by BC026909)
C1orf216 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: UPF0500 protein; UPF0500 protein C1orf216
Gene Aliases: C1orf216
UniProt ID: (Human) Q8TAB5
Entrez Gene ID: (Human) 127703
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support