Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NALRRLMFA VEKSINRKRL WNFTEEEVSE RVTLHICAST KKIFCRTYDI HDPKSSARPA DWKYQSGLSS SWLSLECTVH INIHIPLSAT SVSYTLEKNT KNGLTRWAKE IENGVYLING QVKDEDCDLL EGQ (106-237 aa encoded by BC003397)
May play a role in the trafficking of a subset of G-protein coupled receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hODR-4; LAG1-interacting protein; mODR-4; odorant response abnormal 4; olfactory specific odr-4; Protein odr-4 homolog; transactivated by recombinant transforming growth factor beta; Transactivated by transforming growth factor beta protein 1
Gene Aliases: 1810053E15Rik; C1orf27; odr-4; ODR4; TTG1; TTG1A
UniProt ID: (Mouse) Q4PJX1, (Human) Q5SWX8
Entrez Gene ID: (Mouse) 226499, (Human) 54953
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support