Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MHLGIRSARC VLGMEGQQVI LHLPLSQKGP FWTWEPSAPR TLLQVLQDPA LKDLVLTCPT LPWHSLILRP QYEIQAIMHS ELPGQMDARC WGRERGLPLG AALMEDTPTP SLRISWNFRL LPLYYTEVIL FF
May constitute a control point in macrophage inflammatory response, promoting LPS-induced TNF production.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: basement membrane-induced; Induced by contact to basement membrane 1 protein; Protein ICB-1; Protein THEMIS2; Thymocyte-expressed molecule involved in selection protein 2
Gene Aliases: C1orf38; ICB-1; ICB1; THEMIS2
UniProt ID: (Human) Q5TEJ8
Entrez Gene ID: (Human) 9473
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support