Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QEYEDEERLG EDEYYQVVYY YTVTPSYDDF SADFTIDYSI FESEDRLNRL DKDITEAIET TISLETARAD HPKPVTVKPV TTEPSPDLND AVSSLRSPIP LLLSCAFVQV GMYFM
C1orf54 is a protein coding gene. Gene ontology (GO) annotation include extracellular region/cellular component.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ23221; Uncharacterized protein C1orf54
Gene Aliases: C1orf54
UniProt ID: (Human) Q9H5P3
Entrez Gene ID: (Human) 79630
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support