Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QSYHKLMAL DSTIAHLSLR NNNIDDRGAQ LLGQALSTLH SCNRTLVSLN LGFNHIGDEG AGYIADGLRL NRSLLWLSLA HNRIQDKGAL KLAEVLRAFE LTHTEVVERR RLLLEKGTQE RSRSPSSSRH GDSKTDREKS QMVGISNSAL VDKTDKTQTM KTPKGLGKKK EKSWELAKKE EKLGSGQSPT QGTPKKEDAT KAGKGKVTIP EQKPSRAKGI KIGSREKRSI LLESELVVEA TEVVNPLLEP VEHRDGKVFM PGNKVLLHLN LIRNRITEVG LEGFLATVQY QMQFSKAKSA SKGPVGLLWL SLAKNCFAPQ CPAYAIIQEL MLPRDPIKAK LREDEAMAFF P (210-559 aa encoded by BC033790)
LRRC71 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Leucine-rich repeat-containing protein 71; leucine-rich repeat-containing protein C10orf92
Gene Aliases: C1orf92; LRRC71
UniProt ID: (Human) Q8N4P6
Entrez Gene ID: (Human) 149499
Molecular Function:
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support