Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAGPWTFTLL CGLLAATLIQ ATLSPTAVLI LGPKVIKEKL TQELKDHNAT SILQQLPLLS AMREKPAGGI PVLGSLVNTV LKHVIWLKVI TANILQLQVK PSANDQELLV KIPLDMVAGF NTPLVKTIVE FHMTTEAQAT IRMDTSASGP TRLVLSDCAT SHGSLRIQLL HKLSFLVNAL AKQVMNLLVP SLPNLVKNQL CPVIEASFNG MYADLLQLVK VPISLSIDRL EFDLLYPAIK GDTIQLYLGA KLLDSQGKVT KWFNNSAASL TMPTLDNIPF SLIVSQDVVK AAVAAVLSPE EFMVLLDSVL PESAHRLKSS IGLINEKAAD KLGSTQIVKI LTQDTPEFFI DQGHAKVAQL IVLEVFPSSE ALRPLFTLGI EASSEAQFYT KGDQLILNLN NISSDRIQLM NSGIGWFQPD VLKNIITEII HSILLPNQNG KLRSGVPVSL VKALGFEAAE SSLSKDALVL TPASLWKPTS PVSQ
The protein encoded by this gene may be involved in the innate immune response to bacterial exposure in the mouth, nasal cavities, and lungs. The encoded protein is secreted and is a member of the BPI/LBP/PLUNC protein superfamily. This gene is found with other members of the superfamily in a cluster on chromosome 20.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BPI fold containing family B, member 1; BPI fold-containing family B member 1; long palate, lung and nasal epithelium carcinoma associated 1; Long palate, lung and nasal epithelium carcinoma-associated protein 1; VEMSGP; von Ebner minor salivary gland protein
Gene Aliases: BPIFB1; C20orf114; LPLUNC1; UNQ706/PRO1357
UniProt ID: (Human) Q8TDL5
Entrez Gene ID: (Human) 92747
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support