Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MNKKKKPFLG MPAPLGYVPG LGRGATGFTT RSDIGPARDA NDPVDDRHAP PGKRTVGDQM KKNQAADDDD EDLNDTNYDE FNGYAGSLFS SGPYEKDDEE ADAIYAALDK RMDERRKERR EQREKEEIEK YRMERPKIQQ QFSDLKRKLA EVTEEEWLSI PEVGDARNKR QRNPRYEKLT PVPDSFFAKH LQTGENHTSV DPRQTQFGGL NTPYPGGLNT PYPGGMTPGL MTPGTGELDM RKIGQARNTL MDMRLSQVSD SVSGQTVVDP KGYLTDLNSM IPTHGGDIND IKKARLLLKS VRETNPHHPP AWIASARLEE VTGKLQVARN LIMKGTEMCP KSEDVWLEAA RLQPGDTAKA VVAQAVRHLP QSVRIYIRAA ELETDIRAKK RVLRKALEHV PNSVRLWKAA VELEEPEDAR IMLSRAVECC PTSVELWLAL ARLETYENAR KVLNKARENI PTDRHIWITA AKLEEANGNT QMVEKIIDRA ITSLRANGVE INREQWIQDA EECDRAGSVA TCQAVMRAVI GIGIEEEDRK HTWMEDADSC VAHNALECAR AIYAYALQVF PSKKSVWLRA AYFEKNHGTR ESLEALLQRA VAHCPKAEVL WLMGAKSKWL AGDVPAARSI LALAFQANPN SEEIWLAAVK LESENDEYER ARRLLAKARS SAPTARVFMK SVKLEWVQDN IRAAQDLCEE ALRHYEDFPK LWMMKGQIEE QKEMMEKARE AYNQGLKKCP HSTPLWLLLS RLEEKIGQLT RARAILEKSR LKNPKNPGLW LESVRLEYRA GLKNIANTLM AKALQECPNS GILWSEAIFL EARPQRRTKS VDALKKCEHD PHVLLAVAKL FWSQRKITKA REWFHRTVKI DSDLGDAWAF FYKFELQHGT EEQQEEVRKR CESAEPRHGE LWCAVSKDIA NWQKKIGDIL RLVAGRIKNT F
The protein encoded by this gene appears to be involved in pre-mRNA splicing, possibly acting as a bridging factor between U5 and U4/U6 snRNPs in formation of the spliceosome. The encoded protein also can bind androgen receptor, providing a link between transcriptional activation and splicing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: androgen receptor N-terminal domain transactivating protein-1; Androgen receptor N-terminal domain-transactivating protein 1; ANT-1; p102 U5 small nuclear ribonucleoprotein particle-binding protein; Pre-mRNA-processing factor 6; PRP6 homolog; PRP6 pre-mRNA processing factor 6 homolog; putative mitochondrial outer membrane protein import receptor; U5 snRNP-associated 102 kDa protein; U5-102 kDa protein
Gene Aliases: ANT-1; ANT1; C20orf14; hPrp6; Prp6; PRPF6; RP60; SNRNP102; TOM; U5-102K
UniProt ID: (Human) O94906
Entrez Gene ID: (Human) 24148
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support