Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PVSEYCESE NKWSQEKHSP WEGVSDHLAH REPKSQKPFR KMQEEEEESW STSSDLTISI SEDDLILESP EPQPNPGGKM EGEDGIEALK LIHAEQERVA LSTEKNCILQ TLSSPDSEKE SSTNAPTREP GQTPDSDVPR AQVGQHVATL KEHDNSVKEE ATALLRKALT EECGRRSAIH SSESSCSLPS ILNDNSGIKE AKPAVWLNSV PTREQEVSSG CGDKSKKENV AADIPITETE AYQLLKKATL QDNTNQTENR FQKTDASVSH LSGLNIGSGA FETKTANKIA SEASFSSSEG SPLSRHENKK KPVINLKSNA LWDESDDSNS EIEAALRPRN HNTDDSDDFY D (324-673 aa encoded by BC045826)
Centrosomal protein required for establishing a robust mitotic centrosome architecture that can endure the forces that converge on the centrosomes during spindle formation. Required for stabilizing the expanded pericentriolar material around the centriole.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Centrosomal protein kizuna; Polo-like kinase 1 substrate 1
Gene Aliases: C20orf19; HT013; KIZ; Kizuna; NCRNA00153; PLK1S1; RP69
UniProt ID: (Human) Q2M2Z5
Entrez Gene ID: (Human) 55857
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support