Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The function of the C20orf194 gene has not yet been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNAAF9; Dynein axonemal assembly factor 9; uncharacterized protein C20orf194 homolog
Gene Aliases: C20orf194; DNAAF9
UniProt ID: (Human) Q9UFX9
Entrez Gene ID: (Human) 25943, (Rat) 499891
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support