Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSTHVAGLGL DKMKLGNPQS FLDQEEADDQ QLLEPEAWKT YTERRNALRE FLTSDLSPHL LKRHHARMQL LRKCSYYIEV LPKHLALGDQ NPLVLPSALF QLIDPWKFQR MKKVGTAQTK IQLLLLGDLL EQLDHGRAEL DALLRSPDPR PFLADWALVE RRLADVSAVM DSFLTMMVPG RLHVKHRLVS DVSATKIPHI WLMLSTKMPV VFDRKASAAH QDWARLRWFV TIQPATSEQY ELRFRLLDPR TQQECAQCGV IPVAACTFDV RNLLPNRSYK FTIKRAETST LVYEPWRDSL TLHTKPEPLE GPALSHSV
The exact function of C20orf195 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fibronectin type III domain-containing protein 11
Gene Aliases: C20orf195; FNDC11
UniProt ID: (Human) Q9BVV2
Entrez Gene ID: (Human) 79025
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support