Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MWKRSDHQPK IKAEDGPLVG QFEVLGSVPE PAMPHPLELS EFESFPVFQD IRLHIREVGA QLVKKVNAVF QLDITKNGKT ILRWTIDLKN GSGDMYPGPA RLPADTVFTI PESVFMELVL GKMNPQKAFL AGKFKVSGKV LLSWKLERVF KDWAKF
SCP2D1 (SCP2 Sterol Binding Domain Containing 1) is a Protein Coding gene. Diseases associated with SCP2D1 include Corneal Dystrophy, Posterior Polymorphous, 1 and D-Bifunctional Protein Deficiency. An important paralog of this gene is HSD17B4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SCP2 sterol-binding domain containing 1; SCP2 sterol-binding domain-containing protein 1; sterol carrier protein 2-like protein
Gene Aliases: C20orf79; dJ1068E13.2; HSD22; SCP2D1
UniProt ID: (Human) Q9UJQ7
Entrez Gene ID: (Human) 140856
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support