Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MFRKARRVN VRKRNDSEEE ERERDEEQEP PPLLPPPGTG EEAGPGGGDR APGGESLLGP GPSPPSALTP GLGAEAGGGF PGGAEPGNGL KPRKRPRENK EVPRASLLSF QDEEEENEEV FKVKKSSYSK KIVKLLKKEY KEDLEKSKIK TELNSSAESE QPLDKTGHVK DTNQEDGVII SEHGEDEMDM ESEKEEEKPK TGGAFSNALS SLNVLRPGEI PDAAFIHAAR KKRQMARELG DFTPHDNEPG KGRLVREDEN DASDDEDDDE KRRIVFSVKE KSQRQKIAEE IGIEGSDDDA LVTGEQDEEL SRWEQEQIRK GINIPQVQAS QPAEVNMYYQ NTYQTMPYGS S (1-350 aa encoded by BC030539)
This gene encodes a protein that may bind to GC-rich DNA sequences, which suggests its involvement in the regulation of transcription. Alternative splicing of this gene results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: functional spliceosome-associated protein 105; GC-rich sequence DNA binding factor 1; GC-rich sequence DNA-binding factor 1; GC-rich sequence DNA-binding factor candidate; GC-rich sequence DNA-binding factor homolog; PAX3 and PAX7 binding protein 1; PAX3- and PAX7-binding protein 1; PAX3/7BP
Gene Aliases: 1810007M14Rik; BM020; C21orf66; FSAP105; GCFC; GCFC1; Pax3/7bp; PAXBP1
UniProt ID: (Human) Q9Y5B6, (Mouse) P58501
Entrez Gene ID: (Human) 94104, (Mouse) 67367, (Rat) 681004
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support