Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
C21orf7 plays a critical role in the TGF-beta signaling transduction pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MAP3K7 C-terminal-like protein; TAK1-like protein; TAK1-like protein 1; TAK1-like protein 2; TAK1-like protein 4; TGF-beta activated kinase
Gene Aliases: C21orf7; HC21ORF7; MAP3K7CL; TAK1L; TAKL; TAKL-1; TAKL-2; TAKL-4
UniProt ID: (Human) P57077
Entrez Gene ID: (Human) 56911, (Rat) 304131
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support