Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MFVRPESGEQ GPETLAPASG AEIQRFPVPA VEPVPAPGAD SPPGTALELE EAPEPSCRCP GTAQDQPSEE LPDFMAPPVE PPASALELKV WLELEVAERG GQHSSSQQLP HCSQSWAQWK LWRQRPGFAI WAPLPHWRGT SLIQQSSSPA AEGPAATAAG AVCLPAGGAG EQEKEPVSRG SSRSSCSQRR PPPPGMEVCP QLGIWAICP
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Entrez Gene ID: (Human) 408029
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support