Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: coiled-coil domain containing 164; Coiled-coil domain-containing protein 164; Dynein regulatory complex protein 1; dynein regulatory complex subunit 1 homolog
Gene Aliases: b2b1654Clo; C2orf39; CCDC164; CILD21; DRC1; Gm1060
UniProt ID: (Human) Q96MC2, (Rat) Q5XI65, (Mouse) Q3USS3
Entrez Gene ID: (Human) 92749, (Rat) 362712, (Mouse) 381738
Molecular Function:
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support