Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAGDVGGRS CTDSELLLHP ELLSQEFLLL TLEQKNIAVE TDVRVNKDSL TDLYVQHAIP LPQRDLPKNR WGKMMEKKRE QHEIKNETKR SSTVDGLRKR PLIVFDGSST STSIKVKKTE NGDNDRLKPP PQASFTSNAF RKLSNSSSSV SPLILSSNLP VNNKTEHNNN DAKQNHDLTH RKSPSGPVKS PPLSPVGTTP VKLKRAAPKE EAEAMNNLKP PQAKRKIQHV TWP (1-232 aa encoded by BC001310)
The function of this protein has not been specifically defined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ashwin
Gene Aliases: asw; C2orf49
UniProt ID: (Human) Q9BVC5
Entrez Gene ID: (Human) 79074
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support