Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Immunogen sequence: MGSHPTPGLQ RTTSAGYRLP PTRPPASVSP AARGGPMASR GLAGGCQAPQ ALKAQRV
Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 54%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Uncharacterized protein C2orf50
Gene Aliases: C2orf50
UniProt ID: (Human) Q96LR7
Entrez Gene ID: (Human) 130813
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support