Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GQPVEGAWER PGLPGLLACF SWGPWSRRKN QDVAACRSSA QEDFQEPEEE LPLTAIFPNG DCDDLGRGSK ACDGVVHTPA EP
Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 65%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Uncharacterized protein C2orf72
Gene Aliases: C2orf72
UniProt ID: (Human) A6NCS6
Entrez Gene ID: (Human) 257407
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support