Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
C3AR1 is a receptor for the chemotactic and inflammatory peptide anaphylatoxin C3a. The receptor stimulates chemotaxis, granule enzyme release and superoxide anion production. (Uniprot)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: anaphylatoxin C3a receptor; C3a anaphylatoxin chemotactic receptor; C3a receptor; C3a-R; C3AR; Complement component 3a receptor 1
Gene Aliases: AZ3B; C3AR; C3ar1; C3r1; HNFAG09
UniProt ID: (Mouse) O09047
Entrez Gene ID: (Mouse) 12267
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support