Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
EFCAB12 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EF-hand calcium-binding domain-containing protein 12; EF-hand domain-containing protein C3orf25
Gene Aliases: C3orf25; EFCAB12
UniProt ID: (Human) Q6NXP0
Entrez Gene ID: (Human) 90288
Molecular Function: calmodulin-related
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support