Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MYLRRAVSK TLALPLRAPP NPAPLGKDAS LRRMSSNRFP GSSGSNMIYY LVVGVTVSAG GYYAYKTVTS DQAKHTEHKT NLKEKTKAEI HPFQGEKENV AETEKASSEA PEELIVEAEV VDAEESPSAT VVVIKEASAC PGHVEAAPET TAVSAETGPE VTDAAARETT EVNPETTPEV TNAALDEAVT IDNDKDTTKN ETSDEYAELE EENSPAESES SAGDDLQEEA SVGSEAASAQ G (1-240 aa encoded by BC104174)
MGARP plays a role in the trafficking of mitochondria along microtubules. It regulates the kinesin-mediated axonal transport of mitochondria to nerve terminals along microtubules during hypoxia. It participates in the translocation of TRAK2/GRIF1 from the cytoplasm to the mitochondrion. It also plays a role in steroidogenesis through maintenance of mitochondrial abundance and morphology.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CESP-1; Corneal endothelium-specific protein 1; Hypoxia up-regulated mitochondrial movement regulator protein; Mitochondria-localized glutamic acid-rich protein; Ovary-specific acidic protein; Protein MGARP
Gene Aliases: 4930583H14Rik; AI195347; C4orf49; CESP-1; CESP1; GS3582; HUMMR; MGARP; OSAP; Qsap
UniProt ID: (Human) Q8TDB4, (Mouse) Q8VI64
Entrez Gene ID: (Human) 84709, (Mouse) 67749
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support