Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MNSFNYTTPD YGHYDDKDTL DLNTPVDKTS NTLRVPDILA LVIFAVVFLV GVLGNALVVW VTAFEAKRTI NAIWFLNLAV ADFLSCLALP ILFTSIVQHH HWPFGGAACS ILPSLILLNM YASILLLATI SADRFLLVFK PIWCQNFRGA GLAWIACAVA WGLALLLTIP SFLYRVVREE YFPPKVLCGV DYSHDKRRER AVAIVRLVLG FLWPLLTLTI CYTFILLRTW SRRATRSTKT LKVVVAVVAS FFIFWLPYQV TGIMMSFLEP SSPTFLLLNK LDSLCVSFAY INCCINPIIY VVAGQGFQGR LRKSLPSLLR NVLTEESVVR ESKSFTRSTV DTMAQKTQAV
CD88, also known as C5aR1, is a C5a Anaphylatoxin Receptor that mediates the effects of one of the most potent chemotactic agents known. C5AR1 has been implicated in asthma, and its expression has been reported in myeloid blood cells, brain, liver, lung, spleen, heart, kidney, and intestinal tract. CD88 is the receptor for the chemotactic and inflammatory peptide anaphylatoxin C5a. Functionally, CD88 stimulates chemotaxis, granule enzyme release and superoxide anion production. CD88 is also a component of the classical pathway of the complement cascade; it may be involved with tissue injury associated with ischemia or sepsis and it may be a component of the membrane attack complex. Diseases associated with CD88 include Mannose-Binding Lectin Deficiency and Systemic Lupus Erythematosus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C5a anaphylatoxin chemotactic receptor; C5a anaphylatoxin chemotactic receptor 1; C5a anaphylatoxin receptor; C5a ligand; C5a Receptor; C5a-R; CD88; CD88 antigen; complement component 5 receptor 1
Gene Aliases: C5A; C5AR; C5AR1; C5R1; CD88
UniProt ID: (Human) P21730
Entrez Gene ID: (Human) 728
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support