Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MHYGAATHIQ NSRSHGLETV PGHQRLERGA GGETPEFPGC HSPAPPENFG NELLPLSAPL QGLSEGLYPP GRNKTLPAGV LREGAVQFLH RGLCNSNLSS EASARPSGTQ DELHSSRRKT GQTRREGARK HLVCSFRLYP FTVHTVSPGN SHLALYQVFK AVKLCPSETS FFLSRKSLKS SDPWHPPSLS PNSWNRQAGF RAWSSHLISL SLTCSDSQSR RVSSSQQPPL HSLSSHRRAA HVPE
Binds with and transactivates the corticotropin-releasing hormone (CRH) promoter.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dendritic cell nuclear protein 1; Dendritic cell nuclear protein-1; Dendritic cell-associated nuclear protein
Gene Aliases: C5orf20; DCANP1; DCNP1
UniProt ID: (Human) Q8TF63
Entrez Gene ID: (Human) 140947
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support