Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product # 25249-AP is a rabbit polyclonal antibody raised against the full-length of human C6orf130. The MW of this protein is 17 kDa, and this antibody specifically recognizes the 17 kDa protein.
Immunogen sequence: MASSLNEDP EGSRITYVKG DLFACPKTDS LAHCISEDCR MGAGIAVLFK KKFGGVQELL NQQKKSGEVA VLKRDGRYIY YLITKKRASH KPTYENLQKS LEAMKSHCLK NGVTDLSMPR IGCGLDRLQW ENVSAMIEEV FEATDIKITV YTL (1-152 aa encoded by BC011709)
The function of C6orf130 remains largely unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [Protein ADP-ribosylglutamate] hydrolase OARD1; ADP-ribose glycohydrolase OARD1; O-acetyl-ADP-ribose deacetylase 1; O-acetyl-ADP-ribose deacetylase C6orf130; Terminal ADP-ribose protein glycohydrolase 1
Gene Aliases: C6orf130; dJ34B21.3; OARD1; TARG1
UniProt ID: (Human) Q9Y530
Entrez Gene ID: (Human) 221443
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support