Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAVFLQLLPL LLSRAQGNPG ASLDGRPGDR VNLSCGGVSH PIRWVWAPSF PACKGLSKGR RPILWASSSG TPTVPPLQPF VGRLRSLDSG IRRLELLLSA GDSGTFFCKG RHEDESRTVL HVLGDRTYCK APGPTHGSVY PQLLIPLLGA GLVLGLGALG LVWWLHRRLP PQPIRPLPRF ALSPPHSSTC ENRAPEASKG GRAQDSRGPG PGTEPALCGS GPSSPQQAPP AVHSGPC
G6B is a member of the immunoglobulin (Ig) superfamily with 241 amino acids and is a glycosylated, plasma membrane-bound cell surface receptor, but soluble isoforms encoded by some transcript variants have been found in the endoplasmic reticulum and Golgi before being secreted. Multiple transcript variants encoding different isoforms have been reported. RT-PCR, Western blot, and flow cytometric analyses suggest that G6B is usually expressed on platelets and cross linking of G6B inhibits platelet aggregation and activation in a calcium-independent manner by agonists such as ADP and collagen-related peptide. G6B is an inhibitory surface receptor on platelets and may be an antithrombotic drug target. Isoform B is a putative inhibitory receptor. Isoform A may be its activating counterpart. It is known to bind to heparin. G6B is usually expressed in a restricted set of hematopoietic cell lines like K562, Molt4 and Jurkat and not detected in the cell lines like U937, Raji, Tk, HeLa, NKL, NK62 and YT.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DADB-110M10.2; immunoglobulin receptor; Megakaryocyte and platelet inhibitory receptor G6b; Protein G6b
Gene Aliases: C6orf25; G6B; G6B-B; MPIG6B; NG31
UniProt ID: (Human) O95866
Entrez Gene ID: (Human) 80739
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support