Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MALSCTLNRY LLLMAQEHLE FRLPEIKSLL LLFGGQFASS QETYGKSPFW ILSIPSEDIA RNLMKRTVCA KSIFELWGHG QSPEELYSSL KNYPVEKMVP FLHSDSTYKI KIHTFNKTLT QEEKIKRIDA LEFLPFEGKV NLKKPQHVFS VLEDYGLDPN CIPENPHNIY FGRWIADGQR ELIESYSVKK RHFIGNTSMD AGLSFIMANH GKVKENDIVF DPFVGTGGLQ IACAHFGAYV YGTDIDYNTV HGLGKATRKN QKWRGPDENI RANLRQYGLE KYYLDVLVSD ASKPSWRKGT YFDAIITDPP YGIRESTRRT GSQKEIPKGI EKWEKCPESH VPVSLSYHLS DMFLDLLNFA AETLVLGGRL VYWLPVYTPE YTEEMVPWHP CLELVSNCEQ KLSSHTSRRL ITMEKVKKFE NRDQYSHLLS DHFLPYQGHN SFREKYFSGV TKRIAKEEKS TQE
TRMT11 belongs to the methyltransferase superfamily. It is a catalytic subunit of an S-adenosyl-L-methionine-dependent tRNA methyltransferase complex that mediates the methylation of the guanosine nucleotide at position 10 (m2G10) in tRNAs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: tRNA (guanine(10)-N2)-methyltransferase homolog; tRNA guanosine-2'-O-methyltransferase TRM11 homolog
Gene Aliases: C6orf75; MDS024; TRM11; TRMT11; TRMT11-1
UniProt ID: (Human) Q7Z4G4
Entrez Gene ID: (Human) 60487
Molecular Function:
RNA methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support