Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGPGGRVARL LAPLMWRRAV SSVAGSAVGA EPGLRLLAVQ RLPVGAAFCR ACQTPNFVRG LHSEPGLEER AEGTVNEGRP ESDAADHTGP KFDIDMMVSL LRQENARDIC VIQVPPEMRY TDYFVIVSGT STRHLHAMAF YVVKMYKHLK CKRDPHVKIE GKDTDDWLCV DFGSMVIHLM LPETREIYEL EKLWTLRSYD DQLAQIAPET VPEDFILGIE DDTSSVTPVE LKCE
Required for normal mitochondrial ribosome function and mitochondrial translation (PubMed:22238375, PubMed:23171548). May play a role in ribosome biogenesis by preventing premature association of the 28S and 39S ribosomal subunits (Probable). Interacts with mitochondrial ribosomal protein L14 (MRPL14), probably blocking formation of intersubunit bridge B8, preventing association of the 28S and 39S ribosomal subunits (Probable). Addition to isolated mitochondrial ribosomal subunits partially inhibits translation, probably by interfering with the association of the 28S and 39S ribosomal subunits and the formation of functional ribosomes (Probable). May also participate in the assembly and/or regulation of the stability of the large subunit of the mitochondrial ribosome (PubMed:22238376, PubMed:23171548). May function as a ribosomal silencing factor (Probable). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial assembly of ribosomal large subunit protein 1
Gene Aliases: C7orf30; MALSU1; mtRsfA
UniProt ID: (Human) Q96EH3
Entrez Gene ID: (Human) 115416
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support