Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Plays a role in planar mitotic spindle orientation in retinal progenitor cells (RPCs) and promotes the production of symmetric terminal divisions (By similarity). Negatively regulates the mitotic apical cortex localization of GPSM2. Involved also in positive regulation of cell proliferation and tumor cell growth.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2010317E24Rik; p42.3; Protein Ang; protein C9orf140; protein C9orf140 homolog; Suppressor APC domain-containing protein 2; TS/MDEP; Tumor specificity and mitosis phase-dependent expression protein
Gene Aliases: 2010317E24Rik; 6030458L21Rik; AL033337; ang; C9orf140; p42.3; SAPCD2
UniProt ID: (Human) Q86UD0, (Mouse) Q9D818
Entrez Gene ID: (Human) 89958, (Mouse) 72080, (Rat) 680531
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support