Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGFIFSKSM NESMKNQKEF MLMNARLQLE RQLIMQSEMR ERQMAMQIAW SREFLKYFGT FFGLAAISLT AGAIKKKKPA FLVPIVPLSF ILTYQYDLGY GTLLERMKGE AEDILETEKS KLQLPRGMIT FESIEKARKE QSRFFIDK (1-147 aa encoded by BC008212)
The exact function of C9orf46 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5033414D02Rik; Plasminogen receptor (KT); plasminogen receptor, C-terminal lysine transmembrane protein; Plg-R(KT); transmembrane protein C9orf46
Gene Aliases: AD025; C9orf46; MDS030; Plg-R(KT); PLG-RKT; PLGRKT
UniProt ID: (Human) Q9HBL7
Entrez Gene ID: (Human) 55848
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support