Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
C9orf72 may play a role in endosomal trafficking and autophagy. It is known to interact with HNRNPA1, HNRNPA2B1, UBQLN2, and small Rab GTPSases. C9orf72 localizes in the nucleus, cytoplasm, endosome, lysosome, and autophagosome. Mutations in this gene can result in frontotemporal dementia and/or amyotrophic lateral sclerosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Guanine nucleotide exchange factor C9orf72; Guanine nucleotide exchange factor C9orf72 homolog; protein C9orf72 homolog
Gene Aliases: AI840585; ALSFTD; C9orf72; DENND9; DENNL72; FTDALS; FTDALS1
UniProt ID: (Mouse) Q6DFW0, (Human) Q96LT7, (Rat) Q66HC3
Entrez Gene ID: (Mouse) 73205, (Human) 203228, (Rat) 313155
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support