Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MNNLSFSELC CLFCCPPCPG KIASKLAFLP PDPTYTLMCD ESGSRWTLHL SERADWQYSS REKDAIECFM TRTSKGNRIA CMFVRCSPNA KYTLLFSHGN AVDLGQMSSF YIGLGSRINC NIFSYDYSGY GASSGKPTEK NLYADIEAAW LALRTRYGIR PENVIIYGKS IGTVPSVDLA ARYESAAVIL HSPLTSGMRV AFPDTKKTYC FDAFPNIDKI SKITSPVLII HGTEDEVIDF SHGLALFERC QRPVEPLWVE GAGHNDVELY GQYLERLKQF VSQELVNL
ABHD17B is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Abhydrolase domain-containing protein 17B; abhydrolase domain-containing protein FAM108B1; Alpha/beta hydrolase domain-containing protein 17B; family with sequence similarity 108, member B1
Gene Aliases: ABHD17B; C9orf77; CGI-67; FAM108B1
UniProt ID: (Human) Q5VST6
Entrez Gene ID: (Human) 51104
Molecular Function:
serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support