Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Maternal embryonic leucine-zipper kinase (MELK) is a key regulator of survival of stemlike GBM cells in vitro. MELK expression is increased in breast cancer tissue and this increase is also associated with poor patient survival, as predicted for a candidate oncogene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTP-binding protein Parf; Partner of ARF; putative GTP-binding protein Parf; RAB, member of RAS oncogene family-like 6; Rab-like GTP-binding protein 1C; Rab-like protein 1; Rab-like protein 6; RBEL1
Gene Aliases: B230208H17Rik; C9orf86; PARF; pp8875; RABL6; RBEL1; Rbel1a; Rbel1b; RGD1307615
UniProt ID: (Human) Q3YEC7, (Mouse) Q5U3K5
Entrez Gene ID: (Human) 55684, (Mouse) 227624, (Rat) 362084
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support