Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTGHHGWGYG QDDGPSHWHK LYPIAQGDRQ SPINIISSQA VYSPSLQPLE LSYEACMSLS ITNNGHSVQV DFNDSDDRTV VTGGPLEGPY RLKQFHFHWG KKHDVGSEHT VDGKSFPSEL HLVHWNAKKY STFGEAASAP DGLAVVGVFL ETGDEHPSMN RLTDALYMVR FKGTKAQFSC FNPKCLLPAS RHYWTYPGSL TTPPLSESVT WIVLREPICI SERQMGKFRS LLFTSEDDER IHMVNNFRPP QPLKGRVVKA SFRA
Carbonic anhydrases are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. This gene encodes a glycosylphosphatidyl-inositol-anchored membrane isozyme expressed on the luminal surfaces of pulmonary capillaries and proximal renal tubules. Its exact function is not known; however, it may have a role in inherited renal abnormalities of bicarbonate transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CA-VII; Carbonate dehydratase VII; Carbonic anhydrase 7; Carbonic anhydrase VII; carbonic dehydratase VII
Gene Aliases: CA7; CAVII
UniProt ID: (Human) P43166
Entrez Gene ID: (Human) 766
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support