Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Chromatin assembly factor I (CAF1) is a nuclear complex consisting of p50, p60 (CHAF1B), and p150 (CHAF1A) subunits that assembles histone octamers onto replicating DNA in vitro.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAF-1 subunit A; CAF-I 150 kDa subunit; CAF-I p150; Chromatin assembly factor 1 subunit A; chromatin assembly factor 1, subunit A (p150); chromatin assembly factor I (150 kDa); Chromatin assembly factor I p150 subunit; CTB-50L17.7; hp150; MGC71229
Gene Aliases: CAF; CAF-1; CAF1; CAF1B; CAF1P150; CHAF1A; P150
UniProt ID: (Human) Q13111
Entrez Gene ID: (Human) 10036
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support