Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MESMAVATD GGERPGVPAG SGLSASQRRA ELRRRKLLMN SEQRINRIMG FHRPGSGAEE ESQTKSKQQD SDKLNSLSVP SVSKRVVLGD SVSTGTTDQQ GGVAEVKGTQ LGDKLDSFIK PPECSSDVNL ELRQRNRGDL TADSVQRGSR HGLEQYLSRF EEAMKLRKQL ISEKPSQEDG NTTEE (1-184 aa encoded by BC130325)
The immunosuppressant drug cyclosporin A blocks a calcium-dependent signal from the T-cell receptor (TCR) that normally leads to T-cell activation. When bound to cyclophilin B, cyclosporin A binds and inactivates the key signaling intermediate calcineurin. The protein encoded by this gene functions similarly to cyclosporin A, binding to cyclophilin B and acting downstream of the TCR and upstream of calcineurin by causing an influx of calcium. This integral membrane protein appears to be a new participant in the calcium signal transduction pathway, implicating cyclophilin B in calcium signaling, even in the absence of cyclosporin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcium signal-modulating cyclophilin ligand; calcium-modulating cyclophilin ligand; calcium-signal modulating cyclophilin ligand; cyclophilin B-binding protein; Guided entry of tail-anchored proteins factor CAMLG
Gene Aliases: CAML; CAMLG; GET2
UniProt ID: (Human) P49069, (Rat) Q6DGG9
Entrez Gene ID: (Human) 819, (Rat) 81715
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support