Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MANSMNGRN PGGRGGNPRK GRILGIIDAI QDAVGPPKQA AADRRTVEKT WKLMDKVVRL CQNPKLQLKN SPPYILDILP DTYQHLRLIL SKYDDNQKLA QLSENEYFKI YIDSLMKKSK RAIRLFKEGK ERMYEEQSQD RRNLTKLSLI FSHMLAEIKA IFPNGQFQGD NFRITKADAA EFWRKFFGDK TIVPWKVFRQ CLHEVHQISS GLEAMALKST IDLTCNDYIS VFEFDIFTRL FQPWGSILRN WNFLAVTHPG YMAFLTYDEV KARLQKYSTK PGSYIFRLSC TRLGQWAIGY VTGDGNILQT IPHNKPLFQA LIDGSREGFY LYPDGRSYNP DLTGLCEPTP HDH (1-352 aa encoded by BC032851)
This gene encodes a protein of unknown function. The protein contains a bromodomain, a sequence motif often found in transcriptional coactivators, and localizes to the nucleus in testis and several other cell types.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cas-Br-M (murine) ecotropic retroviral transforming sequence b; Cas-Br-M (murine) ectropic retroviral transforming sequence b; Casitas B-lineage lymphoma b; Casitas B-lineage lymphoma proto-oncogene b; Cbl proto-oncogene B, E3 ubiquitin protein ligase; Cbl proto-oncogene, E3 ubiquitin protein ligase B; E3 ubiquitin-protein ligase CBL-B; RING finger protein 56; RING-type E3 ubiquitin transferase CBL-B; SH3-binding protein CBL-B; Signal transduction protein CBL-B
Gene Aliases: AI429560; AI851073; Cbl-b; CBLB; Nbla00127; RNF56
UniProt ID: (Human) Q13191, (Rat) Q8K4S7, (Mouse) Q3TTA7
Entrez Gene ID: (Human) 868, (Pig) 100621690, (Rat) 171136, (Mouse) 208650
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support