Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
All isoforms of CCDC21 can be detected by this antibody.
Immunogen sequence: MESWQKRYD SLQKIVEKQQ QKMDQLRSQV QSLEQEVAQE EGTSQALREE AQRRDSALQQ LRTAVKELSV QNQDLIEKNL TLQEHLRQAQ PGSPPSPDTA QLALELHQEL ASCLQDLQAV (569-687 aa encoded by BC064528)
This gene encodes a protein that belongs to the centrosome-associated family of proteins. The centrosome is a subcellular organelle in the animal cell that functions as a microtubule organizing center and is involved in cell-cycle progression. Alternate splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: centrosomal protein 85kDa; Centrosomal protein of 85 kDa; Cep85; Coiled-coil domain-containing protein 21
Gene Aliases: CCDC21; CEP85
UniProt ID: (Human) Q6P2H3
Entrez Gene ID: (Human) 64793
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support