Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coiled-coil domain-containing protein 39
Gene Aliases: 4921507O14Rik; AI447426; AI844750; b2b1304Clo; b2b1735Clo; b2b2025.1Clo; CCDC39; CILD14; D3Ertd789e; FAP59; RGD1306277
UniProt ID: (Human) Q9UFE4, (Mouse) Q9D5Y1
Entrez Gene ID: (Human) 339829, (Mouse) 51938, (Rat) 310315
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support