Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Contributes to mitotic spindle assembly, maintenance of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: coiled-coil domain containing 5; coiled-coil domain containing 5 (spindle associated); Coiled-coil domain-containing protein 5; Enhancer of invasion-cluster; HAUS augmin-like complex subunit 1; HAUS augmin-like complex, subunit 1; HEI-C; spindle associated
Gene Aliases: BC024400; CCDC5; HAUS1; HEI-C; HEIC; HsT1461
UniProt ID: (Human) Q96CS2, (Rat) Q9R0A8, (Mouse) Q8BHX1
Entrez Gene ID: (Human) 115106, (Rat) 192228, (Mouse) 225745
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support