Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MASSGAGDP LDSKRGEAPF AQRIDPTREK LTPEQLHSMR QAELAQWQKV LPRRRTRNIV TGLGIGALVL AIYGYTFYSI SQERFLDELE DEAKAARARA LARASGS (1-106 aa encoded by BC002698)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: coiled-coil domain containing 56; Coiled-coil domain-containing protein 56; Cytochrome c oxidase assembly factor 3 homolog, mitochondrial; cytochrome c oxidase assembly protein 3 homolog, mitochondrial; Mitochondrial translation regulation assembly intermediate of cytochrome c oxidase protein of 12 kDa
Gene Aliases: 1810033A19Rik; CCDC56; COA3; D11Ertd99e; HSPC009; MITRAC12; RGD1564337
UniProt ID: (Human) Q9Y2R0, (Mouse) Q9D2R6
Entrez Gene ID: (Human) 28958, (Rat) 498000, (Mouse) 52469
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support