Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MQKTRNTVNT SLVGKQKPHK KHITAENMKS SLVCLTQDQL QQILMTVNQG NRSLSLTENG KEAKSQYSLY LNSISNQPKD ENIMGLFKKT EMVSSVPAEN KSVLNEHQET SKQCEQKIAI ENEWKPADIF STLGERECDR SSLEAKKAQW RKELDEQVAL KKKEKEVSEK WNDPWKKSES DKIIWEKHQI LDQSRETVLL EHPFSAVKQE LQRKWIEELN KQIEDDRQRK IEEKIIYSKG EEHDRWAMHF DSLKSYPGSQ SQLFSRSTHK QPEYFCVSPD TQELADVSSV CTPTTGSQVE PSEEEHIAKP IKDVVMANSK KTNFLRSMTA LLDPAQIEER DRQRQKQLEH QKAITAQVEE KRRKKQLEEE QRKKEEQEEE LRLAQEREEM QKQYEEDILK QKQKEEIMTL KTNELFQTMQ RAQELAQRLK QEQRIRELAQ KGHDTSRLIK NLGVDTIQME YNASNISNSR HDSDEISGKM NTYMNSTTSK KDTGVQTDDL NIGIFTNAES HCGSLMERDI TNCSSPEISA ELIGQFSTKK NKQELTQDKG ASLEKENNRC NDQCNQFTRI EKQTKHMKKY PKRPDWNINK PPKRYIPASE KYPKQLQKQR EEKKVRRQME LLHLVEKNNP GHLSQNRGIS PEIFHSSHQE TESKLRWHLV KKEEEPLNIH SFSKERSPSS PVPVVKNRTQ QTQNTLHLPL KNSSYERENL ISGSNQTELS SGISESSHFI PYVRTNEIYY LDPDAPLSGP STQDPQYQNS QDCGQKRQLF DSDCVRDPLL NPNMVKNRDR QQAILKGLSE LRQGLLQKQK ELESSLLPLA ENQEESFGSS F
Microtubule-binding protein required for ciliogenesis (PubMed:28235840). May function in ciliogenesis by mediating the transport of proteins like BBS4 to the cilium, but also through the organization of the centriolar satellites (PubMed:28235840). Plays a role in retina morphogenesis and/or homeostasis. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coiled-coil domain-containing protein 66; DKFZp686C0433
Gene Aliases: CCDC66
UniProt ID: (Human) A2RUB6
Entrez Gene ID: (Human) 285331
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support