Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCDC98, also known as Abraxas 1, was identified through protein binding studies using the breast and ovarian predisposition protein BRCA1 as the binding target. CCDC98 recruits RAP80, a ubiquitin-binding protein, to BRCA1, allowing the formation of BRCA1 foci in response to DNA damage caused by ionizing radiation. Both CCDC98 and RAP80 are required for DNA damage resistance, G2-M checkpoint control, and DNA repair. Cells depleted of either CCDC98 or RAP80 exhibited increased sensitivity to ionizing radiation, although not as much as in BRCA1-depleted cells, suggesting that CCDC98 and RAP80 control only part of the DNA damage response role of BRCA1. At least two isoforms of CCDC98 are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BRCA1-A complex subunit Abraxas; BRCA1-A complex subunit Abraxas 1; coiled-coil domain containing 98; Coiled-coil domain-containing protein 98; FLJ11520; FLJ12642; FLJ13614; Protein FAM175A
Gene Aliases: 3830405G04Rik; 5630400M01Rik; Abra1; ABRAXAS1; AI506069; AL024423; AV118690; Ccdc98; Fam175a; RGD1305287
UniProt ID: (Mouse) Q8BPZ8, (Rat) Q5I0F1
Entrez Gene ID: (Mouse) 70681, (Rat) 289468
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support