Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: NDAETELMMS KLPLENPVVL NSFHFAADCC TSYISQSIPC SLMKSYFETS SECSKPGVIF LTKKGRQVCA KPSGPGVQDC MKKLKPYSI
This gene is located in a cluster of similar genes in the same region of chromosome 17. These genes encode CC cytokines, which are secreted proteins characterized by two adjacent cysteines. The product of this gene is chemotactic for T cells and monocytes, and acts through C-C chemokine receptor type 1 (CCR1). The proprotein is further processed into numerous smaller functional peptides. Naturally-occurring readthrough transcripts occur from this gene into the downstream gene, CCL14 (chemokine (C-C motif) ligand 14).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-C motif chemokine 15; chemokine (C-C motif) ligand 15; Chemokine CC-2; HCC-2; HCC-2Lkn-1; leukotactin 1; Leukotactin-1; LKN-1; Macrophage inflammatory protein 5; MIP-1 delta; MIP-5; MIP-5MIP-1d; Mrp-2b; NCC-3; NCC-3MIP-1D; NCC3MRP-2B; new CC chemokine 3; SCYA15member 15; small inducible cytokine subfamily A (Cys-Cys), member 15; Small-inducible cytokine A15
Gene Aliases: CCL15; HCC-2; HMRP-2B; LKN-1; LKN1; MIP-1 delta; MIP-1D; MIP-5; MIP5; MRP-2B; NCC-3; NCC3; SCYA15; SCYL3; SY15
UniProt ID: (Human) Q16663
Entrez Gene ID: (Human) 6359
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support