Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
AA Sequence of recombinant protein: discontinued.
Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Exodus-2 (CCL21). Anti-Human Exodus-2 (CCL21)-specific antibody was purified by affinity chromatography and then biotinylated.
Sandwich ELISA: To detect Human Exodus-2 (CCL21) by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25-1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human Exodus-2 (CCL21) (500-P109) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Exodus-2 (CCL21).
Western Blot: To detect hExodus-2 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hExodus-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
500-P109BT-1MG will be provided as 2 x 500 µg
This gene is one of several CC cytokine genes clustered on the p-arm of chromosome 9. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. Similar to other chemokines the protein encoded by this gene inhibits hemopoiesis and stimulates chemotaxis. This protein is chemotactic in vitro for thymocytes and activated T cells, but not for B cells, macrophages, or neutrophils. The cytokine encoded by this gene may also play a role in mediating homing of lymphocytes to secondary lymphoid organs. It is a high affinity functional ligand for chemokine receptor 7 (CCR7) that is expressed on T and B lymphocytes and a known receptor for another member of the cytokine family (small inducible cytokine A19).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 6 Ckine; 6Ckine; beta chemokine exodus-2; Beta-chemokine exodus-2; C-C motif chemokine 21; chemokine (C-C motif) ligand 21; Efficient Chemoattractant for Lymphocytes; secondary lymphoid tissue chemokine; Secondary lymphoid-tissue chemokine; SLC; small inducible cytokine subfamily A (Cys-Cys), member 21; Small-inducible cytokine A21
Gene Aliases: 6Ckine; CCL21; CKb9; ECL; SCYA21; SLC; TCA4; UNQ784/PRO1600
UniProt ID: (Human) O00585
Entrez Gene ID: (Human) 6366
Molecular Function:
cytokine
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support