Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In Western blots, shows 100% cross-reactivity with recombinant human (rh) CCL24. Under reducing conditions, this antibody shows approximately 10-50% cross-reactivity with rhCCL2, 8, 13, 28, and rmCCL2. This antibody does not cross-react with rhCCL1, 3, 4, 5, 7, 9, 11, 14a, 15, 17, 18, 19, 20, 22, 23, 25, rmCCL1, 3, 4, 6, 7, 9, 11, 12, 19, 20, 21, 22, 24, 25, 28, or rrCCL20.
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene is one of several CC cytokine genes clustered on the p-arm of chromosome 9. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. Similar to other chemokines the protein encoded by this gene inhibits hemopoiesis and stimulates chemotaxis. This protein is chemotactic in vitro for thymocytes and activated T cells, but not for B cells, macrophages, or neutrophils. The cytokine encoded by this gene may also play a role in mediating homing of lymphocytes to secondary lymphoid organs. It is a high affinity functional ligand for chemokine receptor 7 (CCR7) that is expressed on T and B lymphocytes and a known receptor for another member of the cytokine family (small inducible cytokine A19).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 6 Ckine; 6Ckine; beta chemokine exodus-2; Beta-chemokine exodus-2; C-C motif chemokine 21; chemokine (C-C motif) ligand 21; Efficient Chemoattractant for Lymphocytes; secondary lymphoid tissue chemokine; Secondary lymphoid-tissue chemokine; SLC; small inducible cytokine subfamily A (Cys-Cys), member 21; Small-inducible cytokine A21
Gene Aliases: 6Ckine; CCL21; CKb9; ECL; SCYA21; SLC; TCA4; UNQ784/PRO1600
UniProt ID: (Human) O00585
Entrez Gene ID: (Human) 6366
Molecular Function:
cytokine
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support