Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Eotaxin 2 belongs to the subfamily of small cytokine CC genes. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity on resting T lymphocytes, a minimal activity on neutrophils, and is negative on monocytes and activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-C motif chemokine 24; CC chemokine CCL24; chemokine (C-C motif) ligand 24; CK-beta-6; Eosinophil chemotactic protein 2; Eotaxin-2; Eotaxin2; MPIF-2; Myeloid progenitor inhibitory factor 2; small inducible cytokine A24; small inducible cytokine subfamily A (Cys-Cys), member 24; Small-inducible cytokine A24
Gene Aliases: CCL24; CKb-6; MPIF-2; MPIF2; SCYA24
UniProt ID: (Human) O00175, (Mouse) Q9JKC0
Entrez Gene ID: (Human) 6369, (Mouse) 56221
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support