Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
AA Sequence of recombinant protein: APYGADTPTA CCFSYGRQIP RKFIADYFET SSLCSQPGVI FLTKRNRQIC ADPKETWVQE YITELELNA.
Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Rat MIP-1alpha (CCL3). Anti-Rat MIP-1alpha (CCL3)-specific antibody was purified by affinity chromatography employing an immobilized Rat MIP-1alpha (CCL3) matrix.
Sandwich ELISA: To detect Rat MIP-1alpha (CCL3) by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.5-2.0 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Rat MIP-1alpha (CCL3) (500-P77Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat MIP-1alpha (CCL3).
Western Blot: To detect Rat MIP-1alpha by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Rat MIP-1alpha is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Both MIP-1alpha and MIP-1beta are structurally and functionally related CC chemokines. They participate in host response to invading bacterial, viral, parasite and fungal pathogens by regulating the trafficking and activation state of selected subgroups of inflammatory cells (e.g. macrophages, lymphocytes and NK cells). While both MIP-1alpha and MIP-1beta exert similar effects on monocytes, their effect on lymphocytes differ; with MIP-1alpha selectively attracting CD8+ lymphocytes, and MIP-1beta selectively attracting CD4+ lymphocytes. Additionally, MIP-1alpha and MIP-1beta have also been shown to be potent chemoattractants for B cells, eosinophils and dendritic cells. Both human and murine MIP-1alpha and MIP-1beta are active on human and murine hematopoietic cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-C motif chemokine 3; H-MIP-1-alpha; LD78-alpha; Macrophage inflammatory protein 1 alpha (Small inducible cytokine A3); Macrophage inflammatory protein 1-alpha; MIP-1-alpha; mip1 alpha; RP23-320E6.7; small inducible cytokine A3; Small-inducible cytokine A3
Gene Aliases: Ccl3; MIP-1a; Mip1a; Scya3
UniProt ID: (Rat) P50229
Entrez Gene ID: (Rat) 25542
Molecular Function:
cytokine
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support